Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtf3c3
Entrez Gene ID
98488
Gene Name
general transcription factor IIIC, polypeptide 3
Gene Aliases
AL022818, TF3C-gamma, TFIIIC102
Chromosome Location
Chr.1: 54397581 - 54438971 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033194.2 NP_001028366.1 17 - 18 2581 138
GenBank mRNA AK145040.1 - - 138
AK160997.1 - 17 - 18 2570 138
AK165830.1 - 17 - 18 2582 138
BC141157.1 - 17 - 18 2571 138

View Full Product Details