The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023514.4 | NP_076003.3 | 4 - 5 | 469 | 148 |
GenBank mRNA | AK009082.1 | - | 4 - 5 | 419 | 148 |
AK049465.1 | - | 4 - 5 | 427 | 148 | |
AK144656.1 | - | 4 - 5 | 427 | 148 | |
BC051936.1 | - | 3 - 4 | 162 | 148 | |
BC069980.1 | - | 2 - 3 | 63 | 148 | |
BC083159.1 | - | 3 - 4 | 230 | 148 | |
BC116437.2 | - | 4 - 5 | 419 | 148 |