The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013025.2 | NP_001013043.1 | 10 - 11 | 2375 | 64 |
GenBank mRNA | AK141742.1 | - | 9 - 10 | 1931 | 64 |
BC072608.1 | - | 10 - 11 | 2375 | 64 | |
BC138867.1 | - | 10 - 11 | 2289 | 64 |