Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr78
Entrez Gene ID
242584
Gene Name
WD repeat domain 78
Gene Aliases
BC028975
Chromosome Location
Chr.4: 103034757 - 103114299 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146254.4 NP_666366.4 10 - 11 1603 103
GenBank mRNA AK030997.1 - 3 - 4 3288 103
BC028975.1 - 10 - 11 1528 103

View Full Product Details