Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Satb2
Entrez Gene ID
212712
Gene Name
special AT-rich sequence binding protein 2
Gene Aliases
mKIAA1034
Chromosome Location
Chr.1: 56793981 - 56978681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139146.2 NP_631885.1 7 - 8 1458 85
GenBank mRNA AF319623.1 - 7 - 8 1458 85
AJ606045.1 - 6 - 7 1176 85
AK129270.1 - 6 - 7 2449 85

View Full Product Details