The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029711.1 | NP_083987.1 | 9 - 10 | 1153 | 71 |
GenBank mRNA | AK083263.1 | - | 9 - 10 | 963 | 71 |
AK151513.1 | - | 9 - 10 | 1127 | 71 | |
AK152356.1 | - | 10 - 11 | 978 | 71 | |
AK170061.1 | - | 10 - 11 | 952 | 71 | |
BC099504.1 | - | 8 - 9 | 805 | 71 | |
BC115750.1 | - | 9 - 10 | 951 | 71 |