Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gulp1
Entrez Gene ID
70676
Gene Name
GULP, engulfment adaptor PTB domain containing 1
Gene Aliases
3110030A04Rik, 5730529O06Rik, CED-6, Ced6, GULP, Gulp-2
Chromosome Location
Chr.1: 44551496 - 44796838 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028450.3 NP_082726.2 6 - 7 648 139
GenBank mRNA AK014093.1 - 5 - 6 397 139
AK017798.1 - 6 - 7 626 139
AK028490.1 - 5 - 6 564 139
AK036670.1 - 6 - 7 648 139
BC032154.1 - 6 - 7 819 139
BY713087.1 - 5 - 6 395 139

View Full Product Details