The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133829.2 | NP_598590.2 | 2 - 3 | 1882 | 67 |
NM_178081.4 | NP_835182.1 | 2 - 3 | 1882 | 67 | |
GenBank mRNA | AB247936.1 | - | 1 - 2 | 1497 | 67 |
AK035995.1 | - | 2 - 3 | 1883 | 67 | |
AK046866.1 | - | 2 - 3 | 2026 | 67 | |
AK171946.1 | - | 2 - 3 | 1923 | 67 | |
BC017534.1 | - | 1 - 2 | 991 | 67 | |
BC029324.1 | - | 1 - 2 | 1239 | 67 |