Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr12
Entrez Gene ID
57750
Gene Name
WD repeat domain 12
Gene Aliases
4933402C23Rik, Ytm1, Ytm1p
Chromosome Location
Chr.1: 60076868 - 60098500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199060.1 NP_001185989.1 2 - 3 586 69
NM_001199061.1 NP_001185990.1 2 - 3 318 69
NM_021312.5 NP_067287.1 1 - 2 408 69
GenBank mRNA AB041608.1 - 2 - 3 587 69
AF239765.1 - 1 - 2 64 69
AK007500.1 - 1 - 2 185 69
AK012022.1 - 1 - 2 162 69
AK016611.1 - 2 - 3 262 69
AK031436.1 - 1 - 2 171 69
AK149085.1 - 1 - 2 90 69
AK161299.1 - 2 - 3 331 69
AU080254.1 - 2 - 3 586 69
AY059431.1 - 1 - 2 99 69
BC004748.1 - 1 - 2 242 69
BC052386.1 - 1 - 2 379 69
BC096651.1 - 1 - 2 153 69
CX564842.1 - 1 - 2 409 69

View Full Product Details