Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Raph1
Entrez Gene ID
77300
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Gene Aliases
9430025M21Rik, C730009O10Rik, Lpd
Chromosome Location
Chr.1: 60483185 - 60567074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045513.3 NP_001038978.3 5 - 6 1016 106
GenBank mRNA CB246004.1 - 2 - 3 547 106

View Full Product Details