Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam117b
Entrez Gene ID
72750
Gene Name
family with sequence similarity 117, member B
Gene Aliases
2810425F24Rik, 6330416D14Rik, Als2cr13
Chromosome Location
Chr.1: 59913006 - 59985348 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037725.3 NP_001032814.1 5 - 6 1251 99
GenBank mRNA AK154812.1 - 5 - 6 1253 99
BC080299.1 - 5 - 6 646 99

View Full Product Details