The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026281.2 | NP_080557.2 | 9 - 10 | 1305 | 73 |
GenBank mRNA | AK010720.1 | - | 9 - 10 | 1148 | 73 |
AK154633.1 | - | 9 - 10 | 1305 | 73 | |
AK171111.1 | - | 9 - 10 | 1282 | 73 | |
BC023389.1 | - | 9 - 10 | 1244 | 73 | |
BC049960.1 | - | 7 - 8 | 941 | 73 | |
BC057900.1 | - | 4 - 5 | 407 | 73 | |
DQ104329.1 | - | 9 - 10 | 1253 | 73 |