The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164566.1 | NP_001158038.1 | 4 - 5 | 329 | 86 |
NM_144882.4 | NP_659131.2 | 6 - 7 | 515 | 86 | |
GenBank mRNA | AK031266.1 | - | 6 - 7 | 470 | 86 |
AK032169.1 | - | 6 - 7 | 450 | 86 | |
BC014764.1 | - | 4 - 5 | 329 | 86 | |
BC021879.1 | - | 6 - 7 | 451 | 86 | |
BY069430.1 | - | 4 - 5 | 285 | 86 | |
BY739337.1 | - | 4 - 5 | 292 | 86 | |
EF410080.1 | - | - | 555 | 86 |