The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113367.1 | NP_001106838.1 | 7 - 8 | 800 | 65 |
NM_029267.3 | NP_083543.2 | 7 - 8 | 745 | 65 | |
GenBank mRNA | AF272859.1 | - | 7 - 8 | 608 | 65 |
AK016403.1 | - | 7 - 8 | 800 | 65 |