Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trak2
Entrez Gene ID
70827
Gene Name
trafficking protein, kinesin binding 2
Gene Aliases
2900022D04Rik, 4733401O11Rik, AI480836, AW046271, Als2cr3, CALS-C, GRIF-1, GRIF1, OIP98, mKIAA0549
Chromosome Location
Chr.1: 58900449 - 58973482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172406.3 NP_765994.2 7 - 8 1217 76
GenBank mRNA AK014647.1 - 7 - 8 1166 76
AK036650.1 - 7 - 8 1166 76
BC060681.1 - 6 - 7 934 76

View Full Product Details