The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172626.2 | NP_766214.2 | 5 - 6 | 772 | 77 |
GenBank mRNA | AY461716.1 | - | 5 - 6 | 747 | 77 |
BC054080.1 | - | 1 - 2 | 220 | 77 |