Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hormad1
Entrez Gene ID
67981
Gene Name
HORMA domain containing 1
Gene Aliases
4921522K05Rik, Nohma
Chromosome Location
Chr.3: 95559640 - 95587940 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289534.1 NP_001276463.1 13 - 14 1104 154
NM_001289537.1 NP_001276466.1 13 - 14 1104 154
NM_026489.3 NP_080765.1 13 - 14 1095 154
GenBank mRNA AK016743.1 - 13 - 14 1098 154
AK163995.1 - 13 - 14 1103 154
AY634284.1 - 13 - 14 1103 154
BC051129.1 - 13 - 14 1103 154

View Full Product Details