Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcgr1
Entrez Gene ID
14129
Gene Name
Fc receptor, IgG, high affinity I
Gene Aliases
AI323638, AV092959, CD64, FcgammaRI, IGGHAFC
Chromosome Location
Chr.3: 96282909 - 96293969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010186.5 NP_034316.1 4 - 5 592 71
GenBank mRNA AF143154.1 - 4 - 5 585 71
AF143155.1 - 4 - 5 585 71
AF143156.1 - 4 - 5 585 71
AF143157.1 - 4 - 5 585 71
AF143158.1 - 4 - 5 585 71
AF143159.1 - 4 - 5 585 71
AF143160.1 - 4 - 5 585 71
AF143161.1 - 4 - 5 585 71
AF143162.1 - 4 - 5 585 71
AF143163.1 - 4 - 5 585 71
AF143164.1 - 4 - 5 585 71
AF143165.1 - 4 - 5 585 71
AF143166.1 - 4 - 5 585 71
AF143167.1 - 4 - 5 585 71
AF143168.1 - 4 - 5 585 71
AF143169.1 - 4 - 5 585 71
AF143170.1 - 4 - 5 585 71
AF143171.1 - 4 - 5 585 71
AF143172.1 - 4 - 5 585 71
AF143173.1 - 4 - 5 585 71
AF143174.1 - 4 - 5 585 71
AF143175.1 - 4 - 5 585 71
AF143176.1 - 4 - 5 585 71
AF143177.1 - 4 - 5 585 71
AF143178.1 - 4 - 5 585 71
AF143179.1 - 4 - 5 585 71
AF143180.1 - 4 - 5 585 71
AF143186.1 - 4 - 5 585 71
AF143187.1 - 4 - 5 585 71
AF143188.1 - 4 - 5 585 71
AK033874.1 - 4 - 5 593 71
M31314.1 - 4 - 5 586 71

View Full Product Details