The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027541.4 | NP_081817.2 | 2 - 3 | 629 | 95 |
GenBank mRNA | AK014398.1 | - | 2 - 3 | 237 | 95 |
AK088194.1 | - | 2 - 3 | 630 | 95 | |
AK139852.1 | - | 2 - 3 | 262 | 95 | |
AK167435.1 | - | 2 - 3 | 266 | 95 | |
BC006782.1 | - | 2 - 3 | 214 | 95 |