Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dab1
Entrez Gene ID
13131
Gene Name
disabled 1
Gene Aliases
AI956902, C630028C02Rik, scm, scr, scrambler, yot
Chromosome Location
Chr.4: 103619546 - 104744844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177259.4 NP_796233.2 13 - 14 1886 81
NR_104385.1 14 - 15 2156 81
GenBank mRNA BC138538.1 - 12 - 13 1607 81
BC138539.1 - 12 - 13 1607 81
Y08379.1 - 13 - 14 1834 81

View Full Product Details