Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lrrc19
- Entrez Gene ID
-
100061
- Gene Name
- leucine rich repeat containing 19
- Gene Aliases
-
9130022A01Rik, AI314124, AL022954, AW261791
- Chromosome Location
-
Chr.4: 94636154 - 94650144 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC061228.1 |
- |
2 - 3 |
151 |
85 |
View Full Product Details