Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Katnal2
Entrez Gene ID
71206
Gene Name
katanin p60 subunit A-like 2
Gene Aliases
3110023G01Rik, 4933439B08Rik
Chromosome Location
Chr.18: 76977147 - 77048225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027721.2 NP_081997.1 12 - 13 1175 83
GenBank mRNA AK017114.1 - 12 - 13 1176 83

View Full Product Details