Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdelc1
Entrez Gene ID
72050
Gene Name
KDEL (Lys-Asp-Glu-Leu) containing 1
Gene Aliases
1810049A15Rik, 5730416C13Rik, EP58, Kdel1
Chromosome Location
Chr.1: 44106546 - 44118857 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023645.3 NP_076134.1 2 - 3 660 64
GenBank mRNA AJ404004.1 - 2 - 3 422 64
AK007835.1 - 2 - 3 653 64
AK035391.1 - 2 - 3 653 64
AK035899.1 - 2 - 3 661 64
AK053315.1 - 2 - 3 653 64
AK079701.1 - 2 - 3 653 64
AK170300.1 - 2 - 3 689 64
BC023141.1 - 2 - 3 640 64

View Full Product Details