Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xrcc5
Entrez Gene ID
22596
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 5
Gene Aliases
AI314015, Ku80, Ku86
Chromosome Location
Chr.1: 72307417 - 72394953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009533.2 NP_033559.2 6 - 7 814 84
GenBank mRNA AK010416.1 - 6 - 7 703 84
AK081633.1 - 6 - 7 815 84
AK089944.1 - 6 - 7 811 84
AK165470.1 - 6 - 7 782 84
AK167312.1 - 6 - 7 703 84
AK168913.1 - 6 - 7 703 84
AK169838.1 - 6 - 7 703 84
BC029218.1 - 6 - 7 686 84
BC051660.1 - 6 - 7 754 84

View Full Product Details