Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tex2
Entrez Gene ID
21763
Gene Name
testis expressed gene 2
Gene Aliases
4930568E07Rik, AI553404, Def-5, Taz4
Chromosome Location
Chr.11: 106502139 - 106613418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198292.3 NP_938034.2 8 - 9 2901 85
GenBank mRNA AK129434.1 - 8 - 9 2901 85

View Full Product Details