Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfia
Entrez Gene ID
18027
Gene Name
nuclear factor I/A
Gene Aliases
1110047K16Rik, 9430022M17Rik, CTF, NF1-A, NF1A
Chromosome Location
Chr.4: 97581717 - 98118876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122953.1 NP_001116425.1 1 - 2 372 66
NM_010905.3 NP_035035.1 1 - 2 372 66
GenBank mRNA AF326553.1 - 1 - 2 71 66
AF326554.1 - 1 - 2 71 66
AK052204.1 - 1 - 2 194 66
AK054310.1 - 1 - 2 235 66
AK137731.1 - 1 - 2 235 66
BC075702.1 - 1 - 2 353 66
BC122878.1 - 1 - 2 94 66
BC122880.1 - 1 - 2 94 66
D90172.1 - 1 - 2 302 66
D90176.1 - 1 - 2 125 66

View Full Product Details