The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025613.1 | NP_001020784.1 | 3 - 4 | 498 | 86 |
NM_001025614.1 | NP_001020785.1 | 2 - 3 | 440 | 86 | |
GenBank mRNA | AK044701.1 | - | 3 - 4 | 499 | 86 |
BB596613.2 | - | 3 - 4 | 536 | 86 | |
BC037040.1 | - | 5 - 6 | 484 | 86 | |
BC141397.1 | - | 2 - 3 | 330 | 86 | |
BC141398.1 | - | 2 - 3 | 330 | 86 |