The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025613.1 | NP_001020784.1 | 4 - 5 | 690 | 73 |
NM_001025614.1 | NP_001020785.1 | 3 - 4 | 632 | 73 | |
GenBank mRNA | AK044701.1 | - | 4 - 5 | 691 | 73 |
BC037040.1 | - | 6 - 7 | 676 | 73 | |
BC141397.1 | - | 3 - 4 | 522 | 73 | |
BC141398.1 | - | 3 - 4 | 522 | 73 |