The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177670.1 | NP_001171141.1 | 5 - 6 | 832 | 91 |
GenBank mRNA | AK167675.1 | - | 5 - 6 | 782 | 91 |
AK169724.1 | - | 4 - 5 | 671 | 91 |