Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde4dip
Entrez Gene ID
83679
Gene Name
phosphodiesterase 4D interacting protein (myomegalin)
Gene Aliases
4732458A06Rik, 9430063L05Rik, C87016, D130016K21Rik, D3Bwg1078e, Usmg4, mKIAA0454
Chromosome Location
Chr.3: 97689828 - 97888707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039376.2 NP_001034465.2 43 - 44 7012 64
NM_001289701.1 NP_001276630.1 43 - 44 7063 64
NM_001289702.1 NP_001276631.1 42 - 43 6910 64
GenBank mRNA AA727946.1 - 1 - 2 49 64
AK040750.1 - 4 - 5 650 64
BC141185.1 - 43 - 44 7063 64
BC145504.1 - 42 - 43 6910 64
BM950385.1 - 3 - 4 568 64

View Full Product Details