Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Akr1cl
- Entrez Gene ID
-
70861
- Gene Name
- aldo-keto reductase family 1, member C-like
- Gene Aliases
-
4921521F21Rik, AI451536, AI503553
- Chromosome Location
-
Chr.1: 65013078 - 65032770 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK014937.1 |
- |
1 - 2 |
156 |
74 |
View Full Product Details