The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097644.1 | NP_001091113.1 | 8 - 9 | 639 | 105 |
GenBank mRNA | AK132782.1 | - | - | 994 | 105 |
CN668580.1 | - | 4 - 5 | 357 | 105 |