The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054064.3 | NP_473405.1 | 1 - 2 | 258 | 135 |
GenBank mRNA | AK141330.1 | - | 1 - 2 | 259 | 135 |
BC114207.1 | - | 1 - 2 | 235 | 135 |