The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164167.1 | NP_001157639.1 | 7 - 8 | 1151 | 135 |
NM_001164168.1 | NP_001157640.1 | 7 - 8 | 1167 | 135 | |
NM_001164169.1 | NP_001157641.1 | 7 - 8 | 1151 | 135 | |
NM_001164170.1 | NP_001157642.1 | 7 - 8 | 1167 | 135 | |
NM_008602.4 | NP_032628.3 | 7 - 8 | 1151 | 135 | |
GenBank mRNA | AF039567.1 | - | 6 - 7 | 596 | 135 |
AK014871.1 | - | 7 - 8 | 1142 | 135 | |
AK029716.1 | - | 7 - 8 | 1152 | 135 | |
AK078097.1 | - | 2 - 3 | 219 | 135 | |
AK078813.1 | - | 7 - 8 | 1121 | 135 | |
AK086653.1 | - | 7 - 8 | 1083 | 135 | |
AK149142.1 | - | 8 - 9 | 1189 | 135 | |
BC005596.1 | - | 7 - 8 | 1028 | 135 | |
BC034711.1 | - | 7 - 8 | 1157 | 135 |