The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033281.3 | NP_001028453.1 | 5 - 6 | 1521 | 83 |
GenBank mRNA | AK133910.1 | - | 5 - 6 | 1522 | 83 |
AK135836.1 | - | 2 - 3 | 395 | 83 | |
AK135840.1 | - | 2 - 3 | 395 | 83 | |
BC139295.1 | - | 5 - 6 | 1551 | 83 | |
DQ340564.1 | - | 5 - 6 | 1097 | 83 | |
DQ340565.1 | - | 5 - 6 | 1016 | 83 |