Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtap
Entrez Gene ID
66902
Gene Name
methylthioadenosine phosphorylase
Gene Aliases
1300019I21Rik, MSAP
Chromosome Location
Chr.4: 89137370 - 89181090 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024433.2 NP_077753.1 2 - 3 244 125
GenBank mRNA AB056100.1 - 2 - 3 232 125
AK005064.1 - 2 - 3 191 125
AK011421.1 - 2 - 3 189 125
AK167319.1 - 2 - 3 245 125
BC003858.1 - 2 - 3 188 125

View Full Product Details