Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrp8
Entrez Gene ID
16975
Gene Name
low density lipoprotein receptor-related protein 8, apolipoprotein e receptor
Gene Aliases
4932703M08Rik, AA921429, AI848122, ApoER2, Lr8b
Chromosome Location
Chr.4: 107802259 - 107876840 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080926.1 NP_001074395.1 18 - 19 2727 61
NR_033496.1 19 - 20 3105 61
GenBank mRNA AJ312058.1 - 18 - 19 2573 61
BC127061.1 - 19 - 20 2480 61
D85463.1 - 19 - 20 3103 61

View Full Product Details