Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spire1
Entrez Gene ID
68166
Gene Name
spire homolog 1 (Drosophila)
Gene Aliases
6030430B19Rik, AI415299, AU022898, AW550622, Spir-1
Chromosome Location
Chr.18: 67488209 - 67610902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176832.2 NP_789802.1 3 - 4 253 63
NM_194355.2 NP_919336.1 3 - 4 351 63
GenBank mRNA AK129296.1 - 5 - 6 858 63
BC058669.1 - 3 - 4 351 63
BC094375.1 - 3 - 4 407 63
BU058399.1 - 3 - 4 352 63

View Full Product Details