Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spire1
Entrez Gene ID
68166
Gene Name
spire homolog 1 (Drosophila)
Gene Aliases
6030430B19Rik, AI415299, AU022898, AW550622, Spir-1
Chromosome Location
Chr.18: 67488209 - 67610902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176832.2 NP_789802.1 8 - 9 905 64
NM_194355.2 NP_919336.1 8 - 9 1003 64
GenBank mRNA BC046486.1 - 1 - 2 86 64
BC058669.1 - 9 - 10 1045 64
BC094375.1 - 9 - 10 1098 64

View Full Product Details