Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnal
Entrez Gene ID
14680
Gene Name
guanine nucleotide binding protein, alpha stimulating, olfactory type
Gene Aliases
2610011C15Rik, 9630020G10Rik, Galphaolf, Gna10, Golf
Chromosome Location
Chr.18: 67088298 - 67226792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010307.3 NP_034437.1 10 - 11 1201 105
NM_177137.5 NP_796111.2 10 - 11 1366 105
GenBank mRNA AK158831.1 - 10 - 11 1202 105
AY179169.1 - 10 - 11 935 105
BC078439.1 - 10 - 11 1328 105
U38503.1 - 3 - 4 311 105

View Full Product Details