Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps45
Entrez Gene ID
22365
Gene Name
vacuolar protein sorting 45 (yeast)
Gene Aliases
AI462172, AW554165, mVps45
Chromosome Location
Chr.3: 95999832 - 96058455 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013841.3 NP_038869.1 1 - 2 773 74
GenBank mRNA AK083678.1 - 1 - 2 149 74
AK146190.1 - 1 - 2 175 74
AK168101.1 - 1 - 2 175 74
BC012691.1 - 1 - 2 149 74
BC058528.1 - 1 - 2 165 74
U66865.1 - 1 - 2 788 74

View Full Product Details