Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps45
Entrez Gene ID
22365
Gene Name
vacuolar protein sorting 45 (yeast)
Gene Aliases
AI462172, AW554165, mVps45
Chromosome Location
Chr.3: 95999832 - 96058455 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013841.3 NP_038869.1 6 - 7 1252 79
GenBank mRNA AK083678.1 - 6 - 7 628 79
AK146190.1 - 6 - 7 654 79
AK168101.1 - 6 - 7 654 79
BC012691.1 - 6 - 7 628 79
BC058528.1 - 6 - 7 644 79
U66865.1 - 6 - 7 1267 79

View Full Product Details