Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps45
Entrez Gene ID
22365
Gene Name
vacuolar protein sorting 45 (yeast)
Gene Aliases
AI462172, AW554165, mVps45
Chromosome Location
Chr.3: 95999832 - 96058455 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013841.3 NP_038869.1 9 - 10 1613 98
GenBank mRNA AK146190.1 - 9 - 10 1015 98
AK168101.1 - 9 - 10 1015 98
BC012691.1 - 9 - 10 989 98
BC058528.1 - 9 - 10 1005 98
U66865.1 - 9 - 10 1628 98

View Full Product Details