The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011867.3 | NP_035997.1 | 13 - 14 | 1754 | 70 |
GenBank mRNA | AF167411.1 | - | 13 - 14 | 1755 | 70 |
BC140995.1 | - | 12 - 13 | 1602 | 70 |