Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cers2
Entrez Gene ID
76893
Gene Name
ceramide synthase 2
Gene Aliases
0610013I17Rik, AI225939, Lass2, TRH3
Chromosome Location
Chr.3: 95314805 - 95323588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029789.1 NP_084065.1 6 - 7 621 120
GenBank mRNA AK002626.1 - 6 - 7 588 120
AK050161.1 - 6 - 7 602 120
AK146106.1 - 6 - 7 600 120
AK159325.1 - 6 - 7 600 120
AY029532.1 - 6 - 7 621 120
BC006847.1 - 6 - 7 593 120

View Full Product Details