The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285897.1 | NP_001272826.1 | 1 - 2 | 358 | 92 |
GenBank mRNA | AK077094.1 | - | 1 - 2 | 208 | 92 |