Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a10
Entrez Gene ID
227059
Gene Name
solute carrier family 39 (zinc transporter), member 10
Gene Aliases
2900042E17Rik, 5430433I10, mKIAA1265
Chromosome Location
Chr.1: 46807544 - 46925772 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172653.2 NP_766241.2 6 - 7 1751 76
GenBank mRNA AK049099.1 - 4 - 5 497 76
AK122483.1 - 5 - 6 1185 76
AK136117.1 - 5 - 6 1463 76
BC052880.1 - 6 - 7 1865 76
BC059214.1 - 4 - 5 635 76
BC062918.1 - 6 - 7 1751 76

View Full Product Details