Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmgxb3
Entrez Gene ID
106894
Gene Name
HMG box domain containing 3
Gene Aliases
2510002C16Rik, A630042L21Rik, AI413166, mKIAA0194
Chromosome Location
Chr.18: 61131277 - 61177050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134134.2 NP_598895.2 6 - 7 1516 81
NM_178277.1 NP_840061.1 6 - 7 1515 81
GenBank mRNA AB045322.1 - 6 - 7 1516 81
AK041866.1 - 6 - 7 1494 81
AK129085.1 - 3 - 4 423 81
AK144993.1 - 6 - 7 1500 81
BC058089.1 - 4 - 5 813 81
BC079580.1 - 6 - 7 1463 81

View Full Product Details