The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145493.1 | NP_663468.1 | 2 - 3 | 380 | 81 |
GenBank mRNA | AK044262.1 | - | 2 - 3 | 388 | 81 |
AY264291.1 | - | 2 - 3 | 269 | 81 | |
BC016082.1 | - | 2 - 3 | 380 | 81 | |
BC029011.1 | - | 2 - 3 | 328 | 81 |