The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008313.4 | NP_032339.2 | 5 - 6 | 720 | 77 |
GenBank mRNA | AJ011369.1 | - | 4 - 5 | 510 | 77 |
AK141587.1 | - | 5 - 6 | 664 | 77 | |
AK141625.1 | - | 5 - 6 | 721 | 77 | |
BC132189.1 | - | 4 - 5 | 549 | 77 | |
BC132191.1 | - | 4 - 5 | 549 | 77 | |
BC145159.1 | - | 4 - 5 | 549 | 77 | |
BC145161.1 | - | 4 - 5 | 549 | 77 | |
Y09585.1 | - | 4 - 5 | 510 | 77 | |
Y09587.1 | - | 4 - 5 | 510 | 77 | |
Y09588.1 | - | 4 - 5 | 510 | 77 |